Assay Details
| Gene Symbol: rpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111421.1 | NP_001104891.1 | 309 | 57 | ||
| GenBank mRNA | AF034949.1 | - | 309 | 57 | ||
| AY107738.1 | - | 320 | 57 | |||
| EU952734.1 | - | 322 | 57 | |||
| EU953522.1 | - | 322 | 57 | |||
| EU961125.1 | - | 322 | 57 | |||
More Information
Set Membership: |
|

rpl30
Genomic Map