Assay Details
| Gene Symbol: ss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111422.1 | NP_001104892.1 | 1494 | 64 | ||
| GenBank mRNA | AF036891.2 | - | 1494 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ss1| Gene Symbol: ss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111422.1 | NP_001104892.1 | 1494 | 64 | ||
| GenBank mRNA | AF036891.2 | - | 1494 | 64 | ||
Set Membership: |
|