Assay Details
| Gene Symbol: rca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111451.1 | NP_001104921.1 | 559 | 59 | ||
| GenBank mRNA | AB564720.1 | - | 539 | 59 | ||
| AF084478.3 | - | 559 | 59 | |||
| AF305876.3 | - | 155 | 59 | |||
| BT063690.1 | - | 534 | 59 | |||
| EU959731.1 | - | 583 | 59 | |||
More Information
Set Membership: |
|

rca1
Genomic Map