Assay Details
| Gene Symbol: rpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111549.1 | NP_001105019.1 | 1027 | 67 | ||
| GenBank mRNA | AF322918.1 | - | 1027 | 67 | ||
| BT033627.1 | - | 1067 | 67 | |||
| EU952105.1 | - | 967 | 67 | |||

rpd3
Genomic Map