Assay Details
| Gene Symbol: pdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111582.1 | NP_001105052.1 | 799 | 66 | ||
| GenBank mRNA | AF370005.1 | - | 799 | 66 | ||
| AY106187.1 | - | 853 | 66 | |||
| BT056069.1 | - | 839 | 66 | |||
| EU970907.1 | - | 772 | 66 | |||
More Information
Set Membership: |
|

pdc3
Genomic Map