Assay Details
| Gene Symbol: pdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111582.1 | NP_001105052.1 | 1753 | 81 | ||
| GenBank mRNA | AF370005.1 | - | 1753 | 81 | ||
| BT056069.1 | - | 1793 | 81 | |||
| D14457.1 | - | 443 | 81 | |||
| EU970907.1 | - | 1726 | 81 | |||
| Z21722.1 | - | 443 | 81 | |||
More Information
Set Membership: |
|

pdc3
Genomic Map