Assay Details
| Gene Symbol: sus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111724.1 | NP_001105194.1 | 1580 | 81 | ||
| GenBank mRNA | AY059416.1 | - | 1423 | 81 | ||
| AY124703.1 | - | 1580 | 81 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

sus2| Gene Symbol: sus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111724.1 | NP_001105194.1 | 1580 | 81 | ||
| GenBank mRNA | AY059416.1 | - | 1423 | 81 | ||
| AY124703.1 | - | 1580 | 81 | |||