Assay Details
| Gene Symbol: sus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111724.1 | NP_001105194.1 | 497 | 78 | ||
| GenBank mRNA | AY059416.1 | - | 340 | 78 | ||
| AY104856.1 | - | 340 | 78 | |||
| AY124703.1 | - | 497 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

sus2| Gene Symbol: sus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111724.1 | NP_001105194.1 | 497 | 78 | ||
| GenBank mRNA | AY059416.1 | - | 340 | 78 | ||
| AY104856.1 | - | 340 | 78 | |||
| AY124703.1 | - | 497 | 78 | |||