Assay Details
| Gene Symbol: sus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111724.1 | NP_001105194.1 | 1414 | 118 | ||
| GenBank mRNA | AY059416.1 | - | 1257 | 118 | ||
| AY104856.1 | - | 1257 | 118 | |||
| AY124703.1 | - | 1414 | 118 | |||

sus2
Genomic Map