Assay Details
| Gene Symbol: asn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111997.1 | NP_001105467.1 | 888 | 71 | ||
| GenBank mRNA | BT035878.1 | - | 71 | |||
| FM211451.1 | - | 71 | ||||
| FM212633.1 | - | 71 | ||||
| X82849.1 | - | 888 | 71 | |||
More Information
Set Membership: |
|

asn1
Genomic Map