Assay Details
| Gene Symbol: pdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112175.1 | NP_001105645.1 | 1503 | 65 | ||
| GenBank mRNA | AF370003.1 | - | 1503 | 65 | ||
| BT064355.1 | - | 1447 | 65 | |||
More Information
Set Membership: |
|

pdc2
Genomic Map