Assay Details
| Gene Symbol: SnrK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112203.1 | NP_001105673.1 | 985 | 100 | ||
| GenBank mRNA | AY486125.1 | - | 985 | 100 | ||
| BT040546.1 | - | 1011 | 100 | |||
| BT060618.1 | - | 1011 | 100 | |||
More Information
Set Membership: |
|

SnrK1
Genomic Map