Assay Details
| Gene Symbol: rop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112249.1 | NP_001105719.1 | 608 | 63 | ||
| GenBank mRNA | AF126055.1 | - | 608 | 63 | ||
| AY108669.1 | - | 645 | 63 | |||
More Information
Set Membership: |
|

rop4
Genomic Map