Assay Details
| Gene Symbol: pht1-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112306.1 | NP_001105776.1 | 1217 | 79 | ||
| GenBank mRNA | AJ830010.1 | - | 1182 | 79 | ||
| AY974046.1 | - | 1217 | 79 | |||
More Information
Set Membership: |
|

pht1-6
Genomic Map