Assay Details
Gene Symbol: pdi8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112334.1 | NP_001105804.1 | 1041 | 87 | ||
GenBank mRNA | AY739290.1 | - | 1041 | 87 | ||
BT033408.1 | - | 1044 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
