Assay Details
| Gene Symbol: MSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112428.1 | NP_001105898.1 | 361 | 66 | ||
| GenBank mRNA | AY856370.1 | - | 361 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MSH1| Gene Symbol: MSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112428.1 | NP_001105898.1 | 361 | 66 | ||
| GenBank mRNA | AY856370.1 | - | 361 | 66 | ||
Set Membership: |
|