Assay Details
| Gene Symbol: LOC100280491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153411.1 | NP_001146883.1 | 1024 | 114 | ||
| GenBank mRNA | EU952042.1 | - | 1024 | 114 | ||
More Information
Set Membership: |
|

LOC100280491
Genomic Map