Assay Details
| Gene Symbol: LOC100280963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153883.1 | NP_001147355.1 | 1251 | 95 | ||
| GenBank mRNA | EU954950.1 | - | 1251 | 95 | ||
More Information
Set Membership: |
|

LOC100280963
Genomic Map