Assay Details
Gene Symbol: LOC100282201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155113.1 | NP_001148585.1 | 217 | 55 | ||
GenBank mRNA | BT070147.1 | - | 55 | |||
EU960008.1 | - | 217 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
