Assay Details
| Gene Symbol: LOC100283668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156569.1 | NP_001150041.1 | 156 | 65 | ||
| GenBank mRNA | EU952988.1 | - | 120 | 65 | ||
| EU965483.1 | - | 156 | 65 | |||
More Information
Set Membership: |
|

LOC100283668
Genomic Map