Assay Details
| Gene Symbol: LOC100284112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157009.1 | NP_001150481.1 | 671 | 73 | ||
| GenBank mRNA | BT061701.1 | - | 601 | 73 | ||
| EU967127.1 | - | 671 | 73 | |||
More Information
Set Membership: |
|

LOC100284112
Genomic Map