Assay Details
| Gene Symbol: LOC100284499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157394.1 | NP_001150866.1 | 1434 | 66 | ||
| GenBank mRNA | EU968583.1 | - | 1434 | 66 | ||
More Information
Set Membership: |
|

LOC100284499
Genomic Map