Assay Details
| Gene Symbol: LOC100285234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158128.1 | NP_001151600.1 | 599 | 68 | ||
| GenBank mRNA | EU971368.1 | - | 599 | 68 | ||
More Information
Set Membership: |
|

LOC100285234
Genomic Map