Assay Details
| Gene Symbol: LOC100285728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158619.1 | NP_001152091.1 | 455 | 67 | ||
| GenBank mRNA | EU973631.1 | - | 455 | 67 | ||
More Information
Set Membership: |
|

LOC100285728
Genomic Map