Assay Details
| Gene Symbol: LOC100285914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158803.1 | NP_001152275.1 | 647 | 104 | ||
| GenBank mRNA | EU974622.1 | - | 647 | 104 | ||
More Information
Set Membership: |
|

LOC100285914
Genomic Map