Assay Details
| Gene Symbol: rrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111406.1 | NP_001104876.1 | 369 | 63 | ||
| GenBank mRNA | AF007793.1 | - | 369 | 63 | ||
| AF250050.1 | - | 389 | 63 | |||
| AY109345.1 | - | 389 | 63 | |||
More Information
Set Membership: |
|

rrb1
Genomic Map