Assay Details
Gene Symbol: hda108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111431.1 | NP_001104901.1 | 986 | 75 | ||
GenBank mRNA | AF045473.1 | - | 986 | 75 | ||
AF440226.1 | - | 920 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
