Assay Details
| Gene Symbol: cesa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111485.1 | NP_001104955.1 | 996 | 85 | ||
| GenBank mRNA | AF200529.1 | - | 996 | 85 | ||
| AY110079.1 | - | 996 | 85 | |||
More Information
Set Membership: |
|

cesa5
Genomic Map