Assay Details
| Gene Symbol: ckb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111504.1 | NP_001104974.1 | 625 | 117 | ||
| GenBank mRNA | AF239816.1 | - | 625 | 117 | ||
| BT040828.1 | - | 600 | 117 | |||
More Information
Set Membership: |
|

ckb1
Genomic Map