Assay Details
| Gene Symbol: prpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111534.1 | NP_001105004.1 | 428 | 92 | ||
| GenBank mRNA | AF273767.1 | - | 428 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prpo2| Gene Symbol: prpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111534.1 | NP_001105004.1 | 428 | 92 | ||
| GenBank mRNA | AF273767.1 | - | 428 | 92 | ||
Set Membership: |
|