Assay Details
| Gene Symbol: spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111536.1 | NP_001105006.1 | 1211 | 66 | ||
| GenBank mRNA | AF283564.2 | - | 1211 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spp1| Gene Symbol: spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111536.1 | NP_001105006.1 | 1211 | 66 | ||
| GenBank mRNA | AF283564.2 | - | 1211 | 66 | ||
Set Membership: |
|