Assay Details
| Gene Symbol: tip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111559.1 | NP_001105029.1 | 145 | 87 | ||
| GenBank mRNA | AF326500.1 | - | 145 | 87 | ||
| EU964091.1 | - | 182 | 87 | |||
More Information
Set Membership: |
|

tip2
Genomic Map