Assay Details
| Gene Symbol: gpm823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111568.1 | NP_001105038.1 | 129 | 75 | ||
| GenBank mRNA | AF330035.1 | - | 129 | 75 | ||
| BT061930.1 | - | 132 | 75 | |||
More Information
Set Membership: |
|

gpm823
Genomic Map