Assay Details
| Gene Symbol: ham101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111604.1 | NP_001105074.1 | 1169 | 104 | ||
| GenBank mRNA | AF440220.1 | - | 1169 | 104 | ||
| BT040127.1 | - | 1201 | 104 | |||
More Information
Set Membership: |
|

ham101
Genomic Map