Assay Details
Gene Symbol: pao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111636.1 | NP_001105106.1 | 452 | 63 | ||
GenBank mRNA | AJ002204.1 | - | 452 | 63 | ||
AY107888.1 | - | 459 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
