Assay Details
| Gene Symbol: nfd110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111654.1 | NP_001105124.1 | 1273 | 67 | ||
| GenBank mRNA | AJ244017.1 | - | 1273 | 67 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

nfd110| Gene Symbol: nfd110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111654.1 | NP_001105124.1 | 1273 | 67 | ||
| GenBank mRNA | AJ244017.1 | - | 1273 | 67 | ||