Assay Details
| Gene Symbol: met4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111703.1 | NP_001105173.1 | 964 | 73 | ||
| GenBank mRNA | AY029557.1 | - | 964 | 73 | ||
| BT037723.1 | - | 1044 | 73 | |||
More Information
Set Membership: |
|

met4
Genomic Map