Assay Details
| Gene Symbol: LOC542105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111738.1 | NP_001105208.1 | 176 | 66 | ||
| GenBank mRNA | AY188756.1 | - | 176 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC542105| Gene Symbol: LOC542105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111738.1 | NP_001105208.1 | 176 | 66 | ||
| GenBank mRNA | AY188756.1 | - | 176 | 66 | ||
Set Membership: |
|