Assay Details
| Gene Symbol: frk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111740.1 | NP_001105210.1 | 856 | 85 | ||
| GenBank mRNA | AY197772.1 | - | 856 | 85 | ||
| BT034483.1 | - | 867 | 85 | |||
| BT039818.1 | - | 912 | 85 | |||
More Information
Set Membership: |
|

frk1
Genomic Map