Assay Details
| Gene Symbol: smh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111756.1 | NP_001105226.1 | 317 | 104 | ||
| GenBank mRNA | AY280630.1 | - | 317 | 104 | ||
| BT037297.1 | - | 416 | 104 | |||
| BT086212.1 | - | 884 | 104 | |||
More Information
Set Membership: |
|

smh6
Genomic Map