Assay Details
| Gene Symbol: S6K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111767.1 | NP_001105237.1 | 904 | 91 | ||
| GenBank mRNA | AY389497.1 | - | 904 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

S6K| Gene Symbol: S6K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111767.1 | NP_001105237.1 | 904 | 91 | ||
| GenBank mRNA | AY389497.1 | - | 904 | 91 | ||
Set Membership: |
|