Assay Details
Gene Symbol: mpk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111768.1 | NP_001105238.1 | 703 | 110 | ||
GenBank mRNA | AY425817.1 | - | 703 | 110 | ||
BT060960.1 | - | 735 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
