Assay Details
| Gene Symbol: mpk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111768.1 | NP_001105238.1 | 1673 | 87 | ||
| GenBank mRNA | AY425817.1 | - | 1673 | 87 | ||
| BT060960.1 | - | 1705 | 87 | |||
More Information
Set Membership: |
|

mpk6
Genomic Map