Assay Details
| Gene Symbol: aprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111773.1 | NP_001105243.1 | 547 | 96 | ||
| GenBank mRNA | AY485263.1 | - | 547 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aprt1| Gene Symbol: aprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111773.1 | NP_001105243.1 | 547 | 96 | ||
| GenBank mRNA | AY485263.1 | - | 547 | 96 | ||
Set Membership: |
|