Assay Details
| Gene Symbol: gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111777.1 | NP_001105247.1 | 344 | 66 | ||
| GenBank mRNA | AY505017.1 | - | 344 | 66 | ||
| U37428.1 | - | 458 | 66 | |||
More Information
Set Membership: |
|

gl1
Genomic Map