Assay Details
| Gene Symbol: gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111777.1 | NP_001105247.1 | 1491 | 84 | ||
| GenBank mRNA | AY505017.1 | - | 1491 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gl1| Gene Symbol: gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111777.1 | NP_001105247.1 | 1491 | 84 | ||
| GenBank mRNA | AY505017.1 | - | 1491 | 84 | ||
Set Membership: |
|