Assay Details
| Gene Symbol: TIDP2941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111805.1 | NP_001105275.1 | 253 | 70 | ||
| GenBank mRNA | AY738148.1 | - | 253 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIDP2941| Gene Symbol: TIDP2941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111805.1 | NP_001105275.1 | 253 | 70 | ||
| GenBank mRNA | AY738148.1 | - | 253 | 70 | ||
Set Membership: |
|