Assay Details
| Gene Symbol: cl3114_1(600) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111823.1 | NP_001105293.1 | 5898 | 112 | ||
| GenBank mRNA | AY894697.1 | - | 5898 | 112 | ||
More Information
Set Membership: |
|

cl3114_1(600)
Genomic Map