Assay Details
| Gene Symbol: cat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111840.1 | NP_001105310.1 | 1311 | 70 | ||
| GenBank mRNA | AY107763.2 | - | 480 | 70 | ||
| AY110648.1 | - | 1313 | 70 | |||
| J02976.1 | - | 1311 | 70 | |||
| X54819.1 | - | 1299 | 70 | |||
More Information
Set Membership: |
|

cat2
Genomic Map