Assay Details
| Gene Symbol: sus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111853.1 | NP_001105323.1 | 1340 | 82 | ||
| GenBank mRNA | AY103630.1 | - | 1476 | 82 | ||
| L22296.1 | - | 1340 | 82 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

sus1| Gene Symbol: sus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111853.1 | NP_001105323.1 | 1340 | 82 | ||
| GenBank mRNA | AY103630.1 | - | 1476 | 82 | ||
| L22296.1 | - | 1340 | 82 | |||